Learn More
Tosoh Bioscience LLC TOYOPEARL IEXPAK HP 4X25ML
Supplier:  Tosoh Bioscience LLC 0019817
Encompass
Media, Process Resins; Tosoh; Toyopearl; LabPak sampler for ion exchange chromatography; IEXPAK HP (DEAE-650S, SP-650S, CM-650S, SuperQ-650S), Particle: 35u m; Matrix: polymer; 4 x 25mL
Catalog No. 50-851-603