Learn More
Tosoh Bioscience LLC TOYOPEARL MEGA SP-550EC 250ML
Supplier:  Tosoh Bioscience LLC 0021805
Encompass
Media, Bulk Cation Exchange Resins; For Ion Exchange Chromatography; Tosoh Toyopearl MegaCap II SP-550EC; Maxtrix: Polymer; Container Size: 250mL
Catalog No. 50-851-695