Learn More
Tosoh Bioscience LLC TOYOPEARL SECPAK HMW 3X150ML
Supplier:  Tosoh Bioscience LLC 0019819
Encompass
Media, Process Resins LabPak for Size Exclusion Chromatography; Tosoh Toyopearl SECPAK HMW (HW-55, 65, 75F), 45 micrometer; Matrix: Polymer; 3 x 150mL
Catalog No. 50-851-605