Learn More
Tosoh Bioscience LLC TOYOPEARL SECPAK LMW 3X150ML
Supplier:  Tosoh Bioscience LLC 0019821
Encompass
Media, Process Resins LabPak for Size Exclusion Chromatography; Tosoh Toyopearl SECPAK LMW (HW-40, 50, 55F), 45 micrometer; Matrix: Polymer; 3 x 150mL
Catalog No. 50-851-607