Learn More
Tosoh Bioscience LLC TOYOPEARL SP-550C 250ML
Supplier:  Tosoh Bioscience LLC 0014028
Encompass
Media, Process Resins; Bulk Cation Exchangers; For Ion Exchange Chromatography; Tosoh Toyopearl SP-550C, 100 micrometer; Matrix: Polymer; Container Size: 250mL
Catalog No. 50-851-332