Learn More
Tosoh Bioscience LLC TOYOPEARL SP-650M 100ML 65UM
Supplier:  Tosoh Bioscience LLC 0043202
Encompass_Preferred
Media, Bulk Cation Exchange Resins; For Ion Exchange Chromatography; Tosoh Toyopearl SP-650M, 65 micrometer; Matrix: Polymer; Container Size: 100mL
Catalog No. 50-851-728