Learn More
Tosoh Bioscience LLC TOYOPEARL SUPERBUTYL550C 100ML
Supplier:  Tosoh Bioscience LLC 0019956
Encompass
Media, Process Resins; For Hydrophobic Interaction Chromatography; Tosoh Toyopearl SuperButyl-550C, 100 micrometer; Matrix: Polymer; Container Size: 100mL
Catalog No. 50-851-612