Learn More
Tosoh Bioscience LLC TOYOPEARL SUPERQ-650C 100ML
Supplier:  Tosoh Bioscience LLC 0043275
Encompass
Media, Bulk Anion Exchange Resins; For Ion Exchange Chromatography; Tosoh Toyopearl SuperQ-650C, 100 micrometer; Matrix: Polymer; Container Size: 100mL
Catalog No. 50-851-735