Learn More
Tosoh Bioscience LLC TOYOPEARL SUPERQ-650M 250ML
Supplier:  Tosoh Bioscience LLC 0017227
Encompass
Media, Bulk Anion Exchange Resins; For Ion Exchange Chromatography; Tosoh Toyopearl SuperQ-650M, 65 micrometer; Matrix: Polymer; Container Size: 250mL
Catalog No. 50-851-430