Learn More
Tosoh Bioscience LLC TOYOPEARL SUPERQ-650S 250ML
Supplier:  Tosoh Bioscience LLC 0017223
Encompass
Media, Bulk Anion Exchange Resins; For Ion Exchange Chromatography; Tosoh Toyopearl SuperQ-650S, 35 micrometer; Matrix: Polymer; Container Size: 250mL
Catalog No. 50-851-429