Learn More
Medchemexpress LLC TP-004 | 1454299-21-8 | 99.2% | 10 MG

Supplier: Medchemexpress LLC HY12086810MG
TP-004 is a small-molecule chemical probe that potently and reversibly inhibits methionine aminopeptidase 2 (MetAP2) with an IC50 of 6 nM. It is supplied as a high-purity solid for biochemical and cellular research.
- Potent, reversible MetAP2 inhibition with IC50 = 6 nM.
- High purity suitable for research (≈99.2%).
- Molecular weight 363.34 g·mol⁻¹; formula C17H16F3N5O.
- Provided as a small solid quantity for in vitro and cellular studies.
- Useful as a chemical probe for MetAP2 target validation and pathway studies.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.