Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Tp-004 | 1454299-21-8 | 99.2% | 5 MG
SDP

Catalog No. 5000232929
Encompass_Preferred

TP-004 is a potent, reversible inhibitor of methionine aminopeptidase 2 (MetAP2) with an IC50 of 6 nM, supplied as a light yellow to khaki solid for research use.

  • Potent reversible MetAP2 inhibitor, IC50 6 nM.
  • Molecular weight 363.34 g/mol; formula C17H16F3N5O.
  • High purity, 99.17% (analytical).
  • Soluble in DMSO (~250 mg/mL); may require ultrasonic agitation.
  • Available in small solid quantities including 5 mg.
  • Store powder at -20°C for long-term stability.

Catalog No. 50-002-32929 Supplier Medchemexpress LLC Supplier No. HY1208685MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

TP-004 is a potent, reversible inhibitor of methionine aminopeptidase 2 (MetAP2) with an IC50 of 6 nM, supplied as a light yellow to khaki solid for research use.

  • Potent reversible MetAP2 inhibitor, IC50 6 nM.
  • Molecular weight 363.34 g/mol; formula C17H16F3N5O.
  • High purity, 99.17% (analytical).
  • Soluble in DMSO (~250 mg/mL); may require ultrasonic agitation.
  • Available in small solid quantities including 5 mg.
  • Store powder at -20°C for long-term stability.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.