Learn More
Medchemexpress LLC TP-004 | 1454299-21-8 | 99.2% | 50 MG
SDP
Catalog No. 5000232821
Encompass_Preferred
TP-004 is a small-molecule chemical probe that potently and reversibly inhibits methionine aminopeptidase 2 (MetAP2) with an IC50 of 6 nM. It is supplied as a high-purity research-grade compound for biochemical and cellular studies.
  • Potent, reversible MetAP2 inhibitor (IC50 6 nM).
  • High purity: 99.2%.
  • Molecular weight 363.34 g/mol; formula C17H16F3N5O.
  • Supplied as a 50 mg research pack suitable for assay and probe development.
  • Intended for research use only; not for human or veterinary use.
Catalog No. 50-002-32821 Supplier Medchemexpress LLC Supplier No. HY12086850MG
TP-004 is a small-molecule chemical probe that potently and reversibly inhibits methionine aminopeptidase 2 (MetAP2) with an IC50 of 6 nM. It is supplied as a high-purity research-grade compound for biochemical and cellular studies.
  • Potent, reversible MetAP2 inhibitor (IC50 6 nM).
  • High purity: 99.2%.
  • Molecular weight 363.34 g/mol; formula C17H16F3N5O.
  • Supplied as a 50 mg research pack suitable for assay and probe development.
  • Intended for research use only; not for human or veterinary use.