Learn More
Medchemexpress LLC TP-021 (BCL6-IN-8c) | 2130878-25-8 | 99.1% | 25 MG
SDP
Catalog No. 5000233205
Encompass_Preferred
TP-021 (BCL6-IN-8c) is a chemical probe that inhibits the interaction between B-cell lymphoma 6 (BCL6) and its corepressors. It is a potent, orally active inhibitor with an IC50 of 0.10 μM, supplied as a 25 mg solid for research use.
  • Potent BCL6-corepressor interaction inhibitor with IC50 0.10 μM.
  • High purity, supporting biochemical and cellular studies.
  • Soluble in DMSO at ≥ 50 mg/mL.
  • Powder stable at -20°C for long-term storage; solvent stability varies.
  • Supplied as a 25 mg solid for laboratory research.
Catalog No. 50-002-33205 Supplier Medchemexpress LLC Supplier No. HY11940225MG
TP-021 (BCL6-IN-8c) is a chemical probe that inhibits the interaction between B-cell lymphoma 6 (BCL6) and its corepressors. It is a potent, orally active inhibitor with an IC50 of 0.10 μM, supplied as a 25 mg solid for research use.
  • Potent BCL6-corepressor interaction inhibitor with IC50 0.10 μM.
  • High purity, supporting biochemical and cellular studies.
  • Soluble in DMSO at ≥ 50 mg/mL.
  • Powder stable at -20°C for long-term storage; solvent stability varies.
  • Supplied as a 25 mg solid for laboratory research.