Learn More
Medchemexpress LLC HY-114965 5mg , TP-064 CAS:2080306-20-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1149655MG
Encompass_Preferred
HY-114965 5mg TP-064 CAS: 2080306-20-1 TP-064 is a potent and selective proteinarginine methyltransferase 4 (PRMT4; CARM1) inhibitor (IC50 <10 nM). TP-064 inhibits dimethylation of BAF155 (IC50 of 340 nM) and MED12 (IC50 of 43 nM). TP-064 is inactive against the other family members except for PRMT6 (IC50 of 1.3 μM). TP-064 has anticancer activities. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-1304