Learn More
Selleck Chemical LLC TP-1454-E1328-5MG
SDP
Catalog No. 503137289
Encompass_Preferred
TP-1454 is an activator of PKM2 with AC50 of 10 Nm in biochemical assays TP-1454 modulates tumor-immune responses by destabilizing T-regulatory cells
Catalog No. 50-313-7289 Supplier Selleck Chemical LLC Supplier No. E13285MG
TP-1454 is an activator of PKM2 with AC50 of 10 Nm in biochemical assays TP-1454 modulates tumor-immune responses by destabilizing T-regulatory cells