Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC TP-238 hydrochloride 10mM 1mL | 2415263-05-5 | 1 ML
SDP

Catalog No. 5000229690
Encompass_Preferred

TP-238 hydrochloride is the hydrochloride salt of TP-238, a potent and selective dual CECR2/BPTF probe used for biochemical and cellular studies of epigenetic reader domains. Offered as a 10 mM solution in DMSO and as solids, it is suitable for target validation and assay development.

  • Potent dual CECR2 and BPTF inhibitor with IC50s of 30 nM and 350 nM.
  • Additional inhibitory activity against BRD9 (pIC50 5.9).
  • Available as a 10 mM DMSO solution (1 mL) and as solids (5-500 mg).
  • Typical purity around 98.8% (source contains conflicting purity statements).
  • Molecular weight 495.04 and formula C22H31ClN6O3S.
  • Store at -20°C under inert atmosphere; in solvent store at -80°C up to 6 months.

Catalog No. 50-002-29690 Supplier Medchemexpress LLC Supplier No. HY114205A10MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

TP-238 hydrochloride is the hydrochloride salt of TP-238, a potent and selective dual CECR2/BPTF probe used for biochemical and cellular studies of epigenetic reader domains. Offered as a 10 mM solution in DMSO and as solids, it is suitable for target validation and assay development.

  • Potent dual CECR2 and BPTF inhibitor with IC50s of 30 nM and 350 nM.
  • Additional inhibitory activity against BRD9 (pIC50 5.9).
  • Available as a 10 mM DMSO solution (1 mL) and as solids (5-500 mg).
  • Typical purity around 98.8% (source contains conflicting purity statements).
  • Molecular weight 495.04 and formula C22H31ClN6O3S.
  • Store at -20°C under inert atmosphere; in solvent store at -80°C up to 6 months.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.