Learn More
Medchemexpress LLC HY-101126 10mg , TP-3654 CAS:1361951-15-6 Purity:>98%
SDP
Catalog No. 502260391
Encompass_Preferred
HY-101126 10mg TP-3654 CAS: 1361951-15-6 TP-3654 is a second-generation Pim kinase inhibitor with Ki values of 5 and 42 nM for Pim-1 and Pim-3, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
This item is not returnable. View return policy
Catalog No. 50-226-0391 Supplier Medchemexpress LLC Supplier No. HY10112610MG
HY-101126 10mg TP-3654 CAS: 1361951-15-6 TP-3654 is a second-generation Pim kinase inhibitor with Ki values of 5 and 42 nM for Pim-1 and Pim-3, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.