Learn More
Abcam TP-3654, 5MG
Supplier: Abcam AB2869595MG
MW 418.5 Da, Purity >=98%. A potent second-generation Pim kinase inhibitor with Ki values of 5 and 42 nM for Pim-1 and Pim-3, respectively. Displays anti-cancer activities.
The product is subject to the following: Abcam Restricted Use Statement