Learn More
Medchemexpress LLC TP-5801 TFA | 99.6% | 641.48 | 5 MG
SDP
Catalog No. 5000359982
Encompass_Preferred
TP-5801 TFA is an orally active TNK1 (non-receptor tyrosine kinase) inhibitor with anti-tumor activity.
  • TNK1 inhibitor with an IC50 of 1.40 nM.
  • Inhibits TNK1-driven, BCR-ABL-driven and IL-3-driven Ba/F3 cell growth.
  • Inhibits TNK1-dependent L540 cell growth.
  • Demonstrates efficacy in mouse survival models.
  • Capable of inhibiting localized tumor growth in vivo.
  • For research use only.
Catalog No. 50-003-59982 Supplier Medchemexpress LLC Supplier No. HY147316A5MG
TP-5801 TFA is an orally active TNK1 (non-receptor tyrosine kinase) inhibitor with anti-tumor activity.
  • TNK1 inhibitor with an IC50 of 1.40 nM.
  • Inhibits TNK1-driven, BCR-ABL-driven and IL-3-driven Ba/F3 cell growth.
  • Inhibits TNK1-dependent L540 cell growth.
  • Demonstrates efficacy in mouse survival models.
  • Capable of inhibiting localized tumor growth in vivo.
  • For research use only.