Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC TP0427736 hydrochloride | 2459963-17-6 | 99.2% | 334.85 | C14H11ClN4S2 | 100 MG
SDP

Supplier:  Medchemexpress LLC HY118528A100MG

Encompass_Preferred

TP0427736 hydrochloride is a research-grade small-molecule inhibitor of the TGF-β type I receptor kinase (ALK5) used for biochemical and cell-based studies. It potently inhibits ALK5 kinase activity and reduces Smad2/3 phosphorylation in cellular assays, making it suitable for research on TGF-β/Smad signaling and related applications.

  • Potent ALK5 inhibition, IC50 = 2.72 nM.
  • Cellular activity: inhibits Smad2/3 phosphorylation in A549 cells, IC50 = 8.68 nM.
  • High reported purity (99.22%).
  • Supplied as the hydrochloride salt; available in research pack sizes including 100 mg.
  • Molecular weight 334.85 g·mol-1; formula C14H11ClN4S2.

Catalog No. 50-002-28591


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.