Learn More
Medchemexpress LLC TP0427736 hydrochloride | 2459963-17-6 | 99.2% | 334.85 | C14H11ClN4S2 | 25 MG
SDP
Catalog No. 5000227735
Encompass_Preferred
TP0427736 hydrochloride is a research-grade small-molecule inhibitor of ALK5 (TGF-β type I receptor), provided as the hydrochloride salt with characterized purity, molecular weight, solubility, and storage recommendations. It is intended for biochemical and cellular studies of TGF-β/Smad signaling.
  • High purity (>99%) suitable for research applications.
  • Potent ALK5 inhibition with low-nanomolar IC50 values.
  • Available in milligram pack sizes for assay development and screening.
  • Soluble in water and DMSO with published solubility conditions.
  • Stored under low-temperature conditions to preserve stability.
Catalog No. 50-002-27735 Supplier Medchemexpress LLC Supplier No. HY118528A25MG
TP0427736 hydrochloride is a research-grade small-molecule inhibitor of ALK5 (TGF-β type I receptor), provided as the hydrochloride salt with characterized purity, molecular weight, solubility, and storage recommendations. It is intended for biochemical and cellular studies of TGF-β/Smad signaling.
  • High purity (>99%) suitable for research applications.
  • Potent ALK5 inhibition with low-nanomolar IC50 values.
  • Available in milligram pack sizes for assay development and screening.
  • Soluble in water and DMSO with published solubility conditions.
  • Stored under low-temperature conditions to preserve stability.