Learn More
Medchemexpress LLC TP0427736 hydrochloride | 2459963-17-6 | 99.2% | 334.85 | C14H11ClN4S2 | 5 MG
SDP
Catalog No. 5000227622
Encompass_Preferred
TP0427736 hydrochloride is a potent ALK5 (TGF-β receptor I) kinase inhibitor supplied as the hydrochloride salt for laboratory research use. It has reported biochemical potency (IC50 2.72 nM), a molecular weight of 334.85 g/mol, and a reported purity of 99.22%.
  • Potent ALK5 kinase inhibitor (IC50 2.72 nM).
  • Provided as hydrochloride salt to improve solubility for assays.
  • High reported purity suitable for research applications.
  • Available in small pack sizes for screening and assay development.
  • Molecular weight 334.85 g/mol.
Catalog No. 50-002-27622 Supplier Medchemexpress LLC Supplier No. HY118528A5MG
TP0427736 hydrochloride is a potent ALK5 (TGF-β receptor I) kinase inhibitor supplied as the hydrochloride salt for laboratory research use. It has reported biochemical potency (IC50 2.72 nM), a molecular weight of 334.85 g/mol, and a reported purity of 99.22%.
  • Potent ALK5 kinase inhibitor (IC50 2.72 nM).
  • Provided as hydrochloride salt to improve solubility for assays.
  • High reported purity suitable for research applications.
  • Available in small pack sizes for screening and assay development.
  • Molecular weight 334.85 g/mol.