Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Tp0427736 hydrochloride | 2459963-17-6 | 99.2% | 334.85 | C14H11ClN4S2 | 50 MG
SDP

Supplier:  Medchemexpress LLC HY118528A50MG

Encompass_Preferred

TP0427736 hydrochloride is the hydrochloride salt of a potent ALK5 (TGF-β type I receptor) kinase inhibitor for in vitro and preclinical research. It inhibits ALK5 with low-nanomolar potency and reduces Smad2/3 phosphorylation in cellular assays.

  • Potent ALK5 inhibitor (ALK5 IC50 2.72 nM).
  • Selective over ALK3 (ALK3 IC50 836 nM).
  • Reduces Smad2/3 phosphorylation in A549 cells induced by TGF-β1 (8.68 nM).
  • Molecular weight 334.85.
  • Chemical formula C14H11ClN4S2.
  • Purity 99.22%.
  • Solid supplied as a 50 mg vial for laboratory research.

Catalog No. 50-002-28593


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.