Learn More
Medchemexpress LLC HY-112144 10mg Medchemexpress, TP0463518 CAS:1558021-37-6 Purity:>98%
SDP
Catalog No. 501969038
Encompass_Preferred
Medchemexpress, HY-112144 10mg TP0463518 CAS:1558021-37-6 TP0463518 is a potent hypoxia-inducible factor prolyl hydroxylases (PHDs) inhibitor with a Ki value of 5.3 nM for human PHD2. TP0463518 also inhibits human PHD1/PHD3 with IC50s of 18 and 63 nM as well as monkey PHD2 with an IC50 value of 22 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9038 Supplier Medchemexpress LLC Supplier No. HY11214410MG
Medchemexpress, HY-112144 10mg TP0463518 CAS:1558021-37-6 TP0463518 is a potent hypoxia-inducible factor prolyl hydroxylases (PHDs) inhibitor with a Ki value of 5.3 nM for human PHD2. TP0463518 also inhibits human PHD1/PHD3 with IC50s of 18 and 63 nM as well as monkey PHD2 with an IC50 value of 22 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.