Learn More
Medchemexpress LLC TP0628103 (TFA) | 2865106-78-9 | 99.6% | 1248.68 | 5 MG
SDP
Catalog No. 5000394553
Encompass_Preferred
TP0628103 (Compound 18) TFA is a selective inhibitor of MMP-7, with an IC50 value of 0.17 nM. MMP-7 plays an important role in cancer and fibrosis.
  • Selective inhibitor of MMP-7
  • IC50 value of 0.17 nM
  • Plays an important role in cancer and fibrosis research
  • For research use only
Catalog No. 50-003-94553 Supplier Medchemexpress LLC Supplier No. HY161169A5MG
TP0628103 (Compound 18) TFA is a selective inhibitor of MMP-7, with an IC50 value of 0.17 nM. MMP-7 plays an important role in cancer and fibrosis.
  • Selective inhibitor of MMP-7
  • IC50 value of 0.17 nM
  • Plays an important role in cancer and fibrosis research
  • For research use only