Learn More
Medchemexpress LLC TP4 (Nile tilapia piscidin) | 1429184-62-2 | 97.1% | 2981.56 | 10 MG
SDP
Catalog No. 5000397030
Encompass_Preferred
TP4 (Nile tilapia piscidin) is a piscidin-like antimicrobial peptide. It inhibits multiple gram positive and negative strains (MIC: 0.03-10 μg/mL). TP4 shows hemolytic activities. It enhances immune response, antioxidant activity, and intestinal health against bacterial infections. TP4 also has anti-tumor effect, and induces necrosis by triggering mitochondrial dysfunction in cancer cells.
  • Inhibits gram-positive and gram-negative bacterial strains
  • Exhibits hemolytic activities
  • Enhances immune response and antioxidant activity
  • Promotes intestinal health against bacterial infections
  • Possesses anti-tumor effects
  • Induces necrosis by triggering mitochondrial dysfunction in cancer cells
  • Soluble in water at ≥ 100 mg/mL
  • Stable for 2 years as a powder at -80°C
Catalog No. 50-003-97030 Supplier Medchemexpress LLC Supplier No. HYP555710MG
TP4 (Nile tilapia piscidin) is a piscidin-like antimicrobial peptide. It inhibits multiple gram positive and negative strains (MIC: 0.03-10 μg/mL). TP4 shows hemolytic activities. It enhances immune response, antioxidant activity, and intestinal health against bacterial infections. TP4 also has anti-tumor effect, and induces necrosis by triggering mitochondrial dysfunction in cancer cells.
  • Inhibits gram-positive and gram-negative bacterial strains
  • Exhibits hemolytic activities
  • Enhances immune response and antioxidant activity
  • Promotes intestinal health against bacterial infections
  • Possesses anti-tumor effects
  • Induces necrosis by triggering mitochondrial dysfunction in cancer cells
  • Soluble in water at ≥ 100 mg/mL
  • Stable for 2 years as a powder at -80°C