Learn More
Medchemexpress LLC Tp4 (nile tilapia piscidin) | 1429184-62-2 | 99.8% | 2981.56 | 1 MG
SDP
Catalog No. 5000399099
Encompass_Preferred
TP4 (Nile tilapia piscidin) is an antimicrobial peptide that shows activity against various gram-positive and gram-negative bacterial strains. It exhibits hemolytic properties and is known to enhance immune response, antioxidant activity, and intestinal health when combating bacterial infections. Additionally, it possesses an anti-tumor effect, initiating necrosis in cancer cells by inducing mitochondrial dysfunction.
  • Inhibits gram-positive and gram-negative bacterial strains.
  • Enhances immune response.
  • Improves antioxidant activity.
  • Promotes intestinal health.
  • Induces necrosis in cancer cells.
  • Triggers mitochondrial dysfunction.
Catalog No. 50-003-99099 Supplier Medchemexpress LLC Supplier No. HYP55571MG
TP4 (Nile tilapia piscidin) is an antimicrobial peptide that shows activity against various gram-positive and gram-negative bacterial strains. It exhibits hemolytic properties and is known to enhance immune response, antioxidant activity, and intestinal health when combating bacterial infections. Additionally, it possesses an anti-tumor effect, initiating necrosis in cancer cells by inducing mitochondrial dysfunction.
  • Inhibits gram-positive and gram-negative bacterial strains.
  • Enhances immune response.
  • Improves antioxidant activity.
  • Promotes intestinal health.
  • Induces necrosis in cancer cells.
  • Triggers mitochondrial dysfunction.