Learn More
CLINICAL GUARD Contec TP Series Non Contact Infrared Thermometer
Catalog No. NC1804613
Encompass_Preferred
The Contec TP Series Non Contact Infrared Thermometer is a handheld infrared thermometer designed to provide fast, accurate and clinical grade temperatures in a safe and controlled manner, equipped with a large back-lit LCD screen and a simple, one button operation. Customization modes allow the thermometer to switch between Celsius and Fahrenheit with a breeze and include a memory function that recalls the last 30 readings in addition to including an object mode function, which can be used to check the ambient temperature of a particular object (e.g. temperature of a bottle of milk, bath water, etc) without contact. An option to silence the thermometer is included to ensure that you can take measurements of patients at rest. Powered by two (2) AAA batteries.
This item is not returnable. View return policy
Catalog No. NC1804613 Supplier CLINICAL GUARD Supplier No. TP500
The Contec TP Series Non Contact Infrared Thermometer is a handheld infrared thermometer designed to provide fast, accurate and clinical grade temperatures in a safe and controlled manner, equipped with a large back-lit LCD screen and a simple, one button operation. Customization modes allow the thermometer to switch between Celsius and Fahrenheit with a breeze and include a memory function that recalls the last 30 readings in addition to including an object mode function, which can be used to check the ambient temperature of a particular object (e.g. temperature of a bottle of milk, bath water, etc) without contact. An option to silence the thermometer is included to ensure that you can take measurements of patients at rest. Powered by two (2) AAA batteries.