Learn More
enQuire Bioreagents Calponin-1 Anti-Human Monoclonal [Clone SPM169], Size: 100ug
Supplier:  enQuire Bioreagents 7157RBM10P1
Encompass_Preferred
Product Type: Antibody. Application: WB, IHC. Conjugate/Tag: Unconjugated. Clone: TP53/1799R. Clonality: Monoclonal. Host: Rabbit. Isotype: IgG. Positive Control: MDA-MB-231 Cells. Breast or Colon carcinoma. Cellular Staining: Nuclear. Buffer: 10mM PBS with 0.05% BSA & 0.05% azide. Also available without BSA & Azide. Concentration: 200 ug/ml. Purity: Protein A/G purified from Bioreactor concentrate. Immunogen: Recombinant full-length human TP53 protein. Storage: Short term: Store at 2-4C. Long term, aliquot and store at -20C. MW: 53kDa.Gene ID: 7157.
Catalog No. 50-188-5242