Learn More
Aviva Systems Biology Corp TP53 Antibody - N-terminal region (ARP30312_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP30312_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TP53. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human TP53. Alternate names: LFS1, TRP53, p53ApplicationELISA, WB, CHIP. Species reactivity: Human
Catalog No. 50-014-54152