Learn More
Aviva Systems Biology Corp TP53BP2 Antibody - N-terminal region (ARP38619_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP38619_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TP53BP2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human TP53BP2. Alternate names: BBP, 53BP2, ASPP2, P53BP2, PPP1R13AApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-56731