Learn More
Aviva Systems Biology Corp TP53I13 Antibody - middle region (ARP51703_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP51703_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TP53I13. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human TP53I13. Alternate names: DSCP1ApplicationWB. Species reactivity: Cow, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-60593