Learn More
Aviva Systems Biology Corp TP53I3 Antibody - C-terminal region (ARP64532_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP64532_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TP53I3. It was validated on Western Blot. . Alternate names: PIG3ApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Pig, Rabbit
Catalog No. 50-288-286