Learn More
Aviva Systems Biology Corp TP53INP2 Antibody - C-terminal region (ARP58965_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP58965_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TP53INP2. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the C terminal region of human TP53INP2. Alternate names: C20orf110, DKFZp434B2411, DKFZp434O0827, DOR, FLJ21759, FLJ23500, PINH, dJ1181N3.1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Human, Mouse, Rat
Catalog No. 50-286-544