Learn More
ABclonal Technology TP53RK Rabbit pAb
SDP
Catalog No. 5000108475
Encompass_Preferred
Enables p53 binding activity and protein serine/threonine kinase activity. Involved in protein phosphorylation. Located in cytoplasm and nucleus. Part of EKC/KEOPS complex. Implicated in Galloway-Mowat syndrome 4.
Catalog No. 50-001-08475 Supplier ABclonal Technology Supplier No. A14952500UL
Enables p53 binding activity and protein serine/threonine kinase activity. Involved in protein phosphorylation. Located in cytoplasm and nucleus. Part of EKC/KEOPS complex. Implicated in Galloway-Mowat syndrome 4.