Learn More
Aviva Systems Biology Corp TP73-AS1 ANTIBODY 50UG/50UL
SDP
Supplier:  Aviva Systems Biology Corp ARP57826_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TP73-AS1. It was validated on Western Blot. . Alternate names: MGC138189ApplicationWB. Species reactivity: Human
Catalog No. 50-286-395