Learn More
Medchemexpress LLC HY-101640 1mg Medchemexpress, TPA 023 CAS:252977-51-8 Purity:>98%
SDP
Catalog No. 501932189
Encompass_Preferred
Medchemexpress, HY-101640 1mg TPA 023 CAS:252977-51-8 TPA 023 is a GABAA α2/α3 subtype-selective agonist, with K i of 0.19-0.41 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2189 Supplier Medchemexpress LLC Supplier No. HY1016401MG
Medchemexpress, HY-101640 1mg TPA 023 CAS:252977-51-8 TPA 023 is a GABAA α2/α3 subtype-selective agonist, with K i of 0.19-0.41 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.