Learn More
ACROBiosystems Recombinant Protein;Biotinylated Human TPBG / 5T4 Protein, His Tag, Avi Tag (Avitag™);HEK293;25UG;TPG,TPBG,5T4,M6P1,5T4AG,WAIF1,5T4M6P1,5T4 oncofetal antigen,Trophoblast glycoprotein,5T4 oncofetal trophoblast glycoprotein
Supplier:  ACROBiosystems TPGH82EB25UG
Encompass_Preferred
Recombinant Protein;Biotinylated Human TPBG / 5T4 Protein, His Tag, Avi Tag (Avitag™);25UG;Biotinylated Human TPBG, His Tag, Avi Tag (TPG-H82Eb) is expressed from human 293 cells (HEK293). It contains AA Ser 32 - Ser 355 (Accession # Q13641-1).;This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag. The protein has a calculated MW of 39.0 kDa. The protein migrates as 50-70 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-165-7626