Learn More
Medchemexpress LLC HY-131614 10mg , TPC2-A1-N CAS:136186-07-7 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY13161410MG
Encompass_Preferred
HY-131614 10mg TPC2-A1-N CAS: 136186-07-7 TPC2-A1-N is a powerful and Ca2+-permeable agonist of two pore channel 2 (TPC2), which plays its role by mimicking the physiological actions of NAADP. TPC2-A1-P reproducibly evokes significant Ca2+ responses from TPC2 (EC50=7.8 μM), and the effect can be blocked by several TPC blockers. TPC2-A1-N can be used to probe different functions of TPC2 channels in intact cells. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-0715