Learn More
Selleck Chemical LLC TPCA-1 10mg 507475-17-4 GW683965
SDP
Supplier:  Selleck Chemical LLC S282410MG
Encompass_Preferred
TPCA-1 (GW683965) is an inhibitor of IKK-2 with IC50 of 17.9 nM in a cell-free assay, inhibits NF-?B pathway, exhibits 22-fold selectivity over IKK-1. TPCA-1 is also an inhibitor of STAT3 and enhances apoptosis. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. NC0976849