Learn More
Medchemexpress LLC HY-10074 100mg Medchemexpress, TPCA-1 CAS:507475-17-4 Purity:98%
SDP
Catalog No. 501636820
Encompass_Preferred
Medchemexpress,HY-10074 100mg TPCA-1 CAS:507475-17-4 Formula:C12H10FN3O2S TPCA-1 is a potent and selective inhibitor of IKK-2 with IC50 of 17.9 nM. Purity:98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-163-6820 Supplier Medchemexpress LLC Supplier No. HY10074100MG
Medchemexpress,HY-10074 100mg TPCA-1 CAS:507475-17-4 Formula:C12H10FN3O2S TPCA-1 is a potent and selective inhibitor of IKK-2 with IC50 of 17.9 nM. Purity:98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.