Learn More
Enzo Life Sciences TPCK (1g). CAS: 402-71-1
SDP
Supplier:  Enzo Life Sciences BMLPI1220001
Encompass_Preferred
Irreversible inhibitor of chymotrypsin-like serine proteases such as granzyme D. Inhibits LPS-induced expression of COX-2 in macrophages. Inhibits γ-irradiation-induced apoptosis in myeloid leukemic cells. Alternative name: N alpha-Tosyl-Phe-chloromethylketone. Purity: ≥98%. Long Term Storage: -20°C.
Catalog No. 50-201-1459