Learn More
Worthington Biochemical Corporation TPCK TREAT. TRYPSIN 500MG
SDP
Catalog No. NC9696087
Encompass
A chromatographically purified, diafiltered, lyophilized powder that has been treated with L-(tosylamido-2-phenyl) ethyl chloromethyl ketone (TPCK) to inhibit contaminating chymotryptic activity [Kostka and Carpenter, J. Biol. Chem. 239, 1799 (1964)].
Catalog No. NC9696087 Supplier Worthington Biochemical Corporation Supplier No. LS003741
A chromatographically purified, diafiltered, lyophilized powder that has been treated with L-(tosylamido-2-phenyl) ethyl chloromethyl ketone (TPCK) to inhibit contaminating chymotryptic activity [Kostka and Carpenter, J. Biol. Chem. 239, 1799 (1964)].