Learn More
Aviva Systems Biology Corp TPD52 Antibody - C-terminal region (ARP48271_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP48271_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TPD52. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the C terminal region of human TPD52. Alternate names: D52, N8L, PC-1, PrLZ, hD52ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Zebrafish
Catalog No. 50-285-355