Learn More
Aviva Systems Biology Corp TPD52L3 Antibody - middle region (ARP58727_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP58727_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TPD52L3. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human TPD52L3. Alternate names: MGC26757, MGC45374, NYD-SP25, hD55, NYDSP25ApplicationWB. Species reactivity: Dog, Horse, Human, Rabbit
Catalog No. 50-014-62404