Learn More
Accuflow Systems Saint-Gobain TPE Braided Tubing, 0.125 in ID, 0.355 in OD
Catalog No. NC2606860
Encompass_Preferred
C-Flex TPE braided tubing is a polyester-braided thermoplastic elastomer hose engineered for high-pressure fluid processing in pharmaceutical and biopharmaceutical applications. The reinforced construction improves burst strength and flexibility while remaining compatible with standard sterilization methods.
  • TPE construction with polyester braid reinforcement.
  • 0.125 in inner diameter, 0.355 in outer diameter, 0.115 in wall thickness.
  • Translucent, animal-derived-component-free (ADCF) formulation.
  • Temperature range suitable for sterilization and cold operations.
  • Maximum working pressure 105 psi at 68°F (20°C).
  • Autoclavable and gamma irradiation compatible.
  • Improved bend radius and kink resistance for pump and transfer lines.
  • Offered in standard 25 ft lengths; custom lengths available.
Catalog No. NC2606860 Supplier Accuflow Systems Supplier No. ACC422253
C-Flex TPE braided tubing is a polyester-braided thermoplastic elastomer hose engineered for high-pressure fluid processing in pharmaceutical and biopharmaceutical applications. The reinforced construction improves burst strength and flexibility while remaining compatible with standard sterilization methods.
  • TPE construction with polyester braid reinforcement.
  • 0.125 in inner diameter, 0.355 in outer diameter, 0.115 in wall thickness.
  • Translucent, animal-derived-component-free (ADCF) formulation.
  • Temperature range suitable for sterilization and cold operations.
  • Maximum working pressure 105 psi at 68°F (20°C).
  • Autoclavable and gamma irradiation compatible.
  • Improved bend radius and kink resistance for pump and transfer lines.
  • Offered in standard 25 ft lengths; custom lengths available.