Learn More
Medchemexpress LLC HY-143218 5mg , TPE-MI CAS:1245606-71-6 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1432185MG
Encompass_Preferred
HY-143218 5mg TPE-MI CAS: 1245606-71-6 TPE-MI (Tetraphenylethene maleimide) is a thiol probe for measuring unfolded protein load and proteostasis in cells. TPE-MI can report imbalances in proteostasis in induced pluripotent stem cell models of Huntington disease, as well as cells transfected with mutant Huntington exon 1 before the formation of visible aggregates. TPE-MI also detects protein damage following dihydroartemisinin treatment of the malaria parasitesPlasmodium falciparum . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9517