Learn More
Medchemexpress LLC TPE-PY | 3032077-04-3 | 560.52 | C35H30BrNO | 10 MG
SDP
Catalog No. 5000297535
Encompass_Preferred
TPE-PY is a mitochondria-targeting fluorescent dye that displays aggregation-induced emission (AIE) via restriction of intramolecular rotation. It is cationic and lipophilic, enabling selective staining of mitochondria in live cells for visualization of location, morphology, and abundance. Supplied as a solid with recommended sealed storage away from moisture and light.
  • Mitochondria-targeting fluorescent dye suitable for live-cell imaging.
  • Aggregation-induced emission mechanism increases fluorescence in aggregated state.
  • Cationic and lipophilic properties promote selective mitochondrial staining.
  • Available in small research quantities; 10 MG format for routine experiments.
  • Solid form with storage guidance to preserve stability.
Catalog No. 50-002-97535 Supplier Medchemexpress LLC Supplier No. HYD263910MG
TPE-PY is a mitochondria-targeting fluorescent dye that displays aggregation-induced emission (AIE) via restriction of intramolecular rotation. It is cationic and lipophilic, enabling selective staining of mitochondria in live cells for visualization of location, morphology, and abundance. Supplied as a solid with recommended sealed storage away from moisture and light.
  • Mitochondria-targeting fluorescent dye suitable for live-cell imaging.
  • Aggregation-induced emission mechanism increases fluorescence in aggregated state.
  • Cationic and lipophilic properties promote selective mitochondrial staining.
  • Available in small research quantities; 10 MG format for routine experiments.
  • Solid form with storage guidance to preserve stability.