Learn More
Accuflow Systems TPE Tubing
Catalog No. 502995723
Encompass_Preferred
Purity One™ OneFlex Biopharmaceutical grade TPE tubing has been formulated for demanding and critical fluid transfer applications in Biopharmaceutical processing. OneFlex is an excellent choice for applications that require heat sealing and sterile welding capabilities, and also can be molded for single-use assemblies such as bottle top inserts. Purity One OneFlex is an excellent choice for applications such as sampling, aseptic welding, aseptic sealing, filtration, and sterile filling and transfer. Full validation guide and USP <665> Extractables testing available.
Catalog No. 50-299-5723 Supplier Accuflow Systems Supplier No. ONEFLEX01250250
Purity One™ OneFlex Biopharmaceutical grade TPE tubing has been formulated for demanding and critical fluid transfer applications in Biopharmaceutical processing. OneFlex is an excellent choice for applications that require heat sealing and sterile welding capabilities, and also can be molded for single-use assemblies such as bottle top inserts. Purity One OneFlex is an excellent choice for applications such as sampling, aseptic welding, aseptic sealing, filtration, and sterile filling and transfer. Full validation guide and USP <665> Extractables testing available.